Product Details

SNP ID
rs79587292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49698836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTTCTGCCTGTGAGTTTCCTAT[C/T]TCTATGCACAGGCCTGGCCCCTGGG
Phenotype
MIM: 142408 MIM: 614472
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MST1 PubMed Links

Gene Details

Gene
MST1
Gene Name
macrophage stimulating 1
There are no transcripts associated with this gene.

Gene
RNF123
Gene Name
ring finger protein 123
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022064.4 Intron NP_071347.2
XM_006713285.1 Intron XP_006713348.1
XM_011533995.1 Intron XP_011532297.1
XM_017007018.1 Intron XP_016862507.1

View Full Product Details