Product Details

SNP ID
rs72930617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:100754941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGTTGTCTTTAAAAATCTCCCA[A/T]TTAGTACAAATGACTTGGCAGGTTA
Phenotype
MIM: 606279 MIM: 602498
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ABI3BP PubMed Links

Gene Details

Gene
ABI3BP
Gene Name
ABI family member 3 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015429.3 Intron NP_056244.2
XM_005247281.2 Intron XP_005247338.1
XM_005247282.2 Intron XP_005247339.1
XM_005247283.2 Intron XP_005247340.1
XM_005247284.2 Intron XP_005247341.1
XM_005247285.2 Intron XP_005247342.1
XM_005247287.2 Intron XP_005247344.1
XM_005247288.2 Intron XP_005247345.1
XM_005247289.2 Intron XP_005247346.1
XM_005247290.2 Intron XP_005247347.1
XM_005247291.2 Intron XP_005247348.1
XM_005247293.2 Intron XP_005247350.1
XM_005247294.2 Intron XP_005247351.1
XM_005247295.2 Intron XP_005247352.1
XM_005247296.2 Intron XP_005247353.1
XM_005247298.2 Intron XP_005247355.1
XM_005247299.3 Intron XP_005247356.1
XM_005247300.2 Intron XP_005247357.1
XM_005247301.2 Intron XP_005247358.1
XM_005247302.2 Intron XP_005247359.1
XM_005247303.2 Intron XP_005247360.1
XM_005247305.2 Intron XP_005247362.1
XM_005247306.2 Intron XP_005247363.1
XM_005247309.1 Intron XP_005247366.1
XM_006713569.2 Intron XP_006713632.1
XM_006713570.2 Intron XP_006713633.1
XM_006713571.2 Intron XP_006713634.1
XM_006713572.2 Intron XP_006713635.1
XM_011512639.1 Intron XP_011510941.1
XM_011512640.1 Intron XP_011510942.1
XM_011512641.1 Intron XP_011510943.1
XM_011512643.1 Intron XP_011510945.1
XM_011512644.1 Intron XP_011510946.1
XM_011512645.1 Intron XP_011510947.1
XM_011512646.1 Intron XP_011510948.1
XM_011512647.1 Intron XP_011510949.1
XM_011512648.1 Intron XP_011510950.1
XM_011512649.1 Intron XP_011510951.1
XM_011512651.1 Intron XP_011510953.1
XM_011512652.1 Intron XP_011510954.1
XM_011512653.1 Intron XP_011510955.1
XM_011512654.1 Intron XP_011510956.1
XM_011512656.1 Intron XP_011510958.1
XM_011512657.1 Intron XP_011510959.1
XM_011512658.1 Intron XP_011510960.1
XM_011512659.1 Intron XP_011510961.1
XM_011512660.1 Intron XP_011510962.1
XM_017006105.1 Intron XP_016861594.1
XM_017006106.1 Intron XP_016861595.1
XM_017006107.1 Intron XP_016861596.1
XM_017006108.1 Intron XP_016861597.1
Gene
TFG
Gene Name
TRK-fused gene
There are no transcripts associated with this gene.

View Full Product Details