Product Details

SNP ID
rs9827364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:36987121 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTTTGGGTGATAAGGACATTCT[A/G]CAGGTGTAAAAACAAGTGAACTGAA
Phenotype
MIM: 607911 MIM: 120436
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPM2AIP1 PubMed Links

Gene Details

Gene
EPM2AIP1
Gene Name
EPM2A interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014805.3 6184 UTR 3 NP_055620.1
Gene
MLH1
Gene Name
mutL homolog 1
There are no transcripts associated with this gene.

View Full Product Details