Product Details

SNP ID
rs61739609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:75587785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTGCTGGGACCATAGGAGCTTG[A/G]TTCTTCACAGGCTCAGGAATCCTAG
Phenotype
MIM: 601783
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAP6 PubMed Links

Gene Details

Gene
MAP6
Gene Name
microtubule associated protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033063.1 4368 Silent Mutation AAC,AAT N572N NP_149052.1
NM_207577.1 4368 Intron NP_997460.1
XM_006718556.3 4368 Intron XP_006718619.1
XM_011545039.2 4368 Intron XP_011543341.1
XM_017017755.1 4368 Silent Mutation AAC,AAT N588N XP_016873244.1
XM_017017756.1 4368 Silent Mutation AAC,AAT N287N XP_016873245.1
XM_017017757.1 4368 Silent Mutation AAC,AAT N271N XP_016873246.1
XM_017017758.1 4368 Silent Mutation AAC,AAT N259N XP_016873247.1

View Full Product Details