Product Details

SNP ID
rs73329106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47209754 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGATTGAGTCATAAACCTTTTCC[A/G]TCAAGAATGAGGTGCTGCTTTGAGG
Phenotype
MIM: 160770
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYL4 PubMed Links

Gene Details

Gene
MYL4
Gene Name
myosin light chain 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002841.1 Intron NP_001002841.1
NM_002476.2 Intron NP_002467.1
XM_005257391.4 Intron XP_005257448.1
XM_011524839.2 Intron XP_011523141.2
XM_017024683.1 Intron XP_016880172.1
XM_017024684.1 Intron XP_016880173.1

View Full Product Details