Product Details

SNP ID
rs7251754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7478194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACCCACCAGAGGCTGTGATGAC[A/T]CCCCCATGACCTGAGCACAACAGTC
Phenotype
MIM: 616432 MIM: 607583
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARHGEF18 PubMed Links

Gene Details

Gene
ARHGEF18
Gene Name
Rho/Rac guanine nucleotide exchange factor 18
There are no transcripts associated with this gene.

Gene
LOC100128573
Gene Name
uncharacterized LOC100128573
There are no transcripts associated with this gene.

Gene
PEX11G
Gene Name
peroxisomal biogenesis factor 11 gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270539.1 Intron NP_001257468.1
NM_001300881.1 Intron NP_001287810.1
NM_080662.3 Intron NP_542393.1
XM_005272506.2 Intron XP_005272563.1
XM_011528426.2 Intron XP_011526728.1
XM_011528427.2 Intron XP_011526729.1
XM_011528428.1 Intron XP_011526730.1
XM_011528429.2 Intron XP_011526731.1
XM_011528431.2 Intron XP_011526733.1
XM_011528432.1 Intron XP_011526734.1

View Full Product Details