Product Details

SNP ID
rs144702902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:99401094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATTGGTTCGTCTATCCCTTGAGC[A/G]ACTTCTACTTCTACGTCTCTCTCGG
Phenotype
MIM: 605196 MIM: 616653
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COQ3 PubMed Links

Gene Details

Gene
COQ3
Gene Name
coenzyme Q3, methyltransferase
There are no transcripts associated with this gene.

Gene
PNISR
Gene Name
PNN interacting serine and arginine rich protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322405.1 2026 Missense Mutation NP_001309334.1
NM_001322406.1 2026 Missense Mutation NP_001309335.1
NM_001322408.1 2026 Missense Mutation NP_001309337.1
NM_001322410.1 2026 Missense Mutation NP_001309339.1
NM_001322412.1 2026 Missense Mutation NP_001309341.1
NM_001322413.1 2026 Missense Mutation NP_001309342.1
NM_001322414.1 2026 Missense Mutation NP_001309343.1
NM_001322415.1 2026 Missense Mutation NP_001309344.1
NM_001322416.1 2026 Intron NP_001309345.1
NM_001322417.1 2026 Intron NP_001309346.1
NM_001322418.1 2026 Intron NP_001309347.1
NM_001322419.1 2026 Intron NP_001309348.1
NM_015491.2 2026 Missense Mutation NP_056306.1
NM_032870.3 2026 Missense Mutation NP_116259.2
XM_005266912.3 2026 Missense Mutation XP_005266969.2
XM_017010710.1 2026 Missense Mutation XP_016866199.1
XM_017010711.1 2026 Missense Mutation XP_016866200.1
XM_017010712.1 2026 Missense Mutation XP_016866201.1
XM_017010713.1 2026 Missense Mutation XP_016866202.1

View Full Product Details