Product Details

SNP ID
rs2234911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142613783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGCTGACGGTGTAGTCGTAGCC[G/A]TCTGCCTCGTGGCAGTAGCTCTCGG
Phenotype
MIM: 612461
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ARC PubMed Links

Gene Details

Gene
ARC
Gene Name
activity regulated cytoskeleton associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015193.4 690 Silent Mutation GAC,GAT D163D NP_056008.1

View Full Product Details