Product Details

SNP ID
rs4263535
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:161857823 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGGATTCATTCTTGTCCATAATC[A/G]CTTTATTTGGTGGCAAGGTGTGGGG
Phenotype
MIM: 137160
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GABRA1 PubMed Links

Gene Details

Gene
GABRA1
Gene Name
gamma-aminobutyric acid type A receptor alpha1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000806.5 Intron NP_000797.2
NM_001127643.1 Intron NP_001121115.1
NM_001127644.1 Intron NP_001121116.1
NM_001127645.1 Intron NP_001121117.1
NM_001127648.1 Intron NP_001121120.1

View Full Product Details