Product Details

SNP ID
rs13072
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:440579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACTGACGTATTTCACACTCTCT[A/C]CACACCCAGCATGATTTGTCTGAAC
Phenotype
MIM: 615928
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM101B PubMed Links

Gene Details

Gene
FAM101B
Gene Name
family with sequence similarity 101 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182705.2 3294 UTR 3 NP_874364.1

View Full Product Details