Product Details

SNP ID
rs7488085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:131902684 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAGGCGGCAAGTTGGGACCCACC[C/T]CCCGGGGTGGGGGTGATGGTGCAGG
Phenotype
MIM: 603168
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ULK1 PubMed Links

Gene Details

Gene
ULK1
Gene Name
unc-51 like autophagy activating kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003565.2 Intron NP_003556.1
XM_011538798.2 Intron XP_011537100.1
XM_011538799.2 Intron XP_011537101.1

View Full Product Details