Product Details
- SNP ID
-
rs7238205
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:39207607 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTTATCTTAAGTTACAATGCAAC[A/G]TTAAAATCTAAGTGGAAATGAGATT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR924HG
PubMed Links
Gene Details
- Gene
- MIR924HG
- Gene Name
- MIR924 host gene
There are no transcripts associated with this gene.
View Full Product Details