Product Details

SNP ID
rs4128658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45064388 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCAAGGGCCTAGTCTGGAGGAGAG[A/G]GTCTAATTCTGGCACTAAGAATCTG
Phenotype
MIM: 160993
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCAKD PubMed Links

Gene Details

Gene
DCAKD
Gene Name
dephospho-CoA kinase domain containing
There are no transcripts associated with this gene.

Gene
NMT1
Gene Name
N-myristoyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021079.4 Intron NP_066565.1
XM_011524858.2 Intron XP_011523160.1

View Full Product Details