Product Details
- SNP ID
-
rs6744348
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:27795158 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTGCTGACTCCTTAGAGTTTTAC[A/C]ATCCCTTAAAAGTCCTGTGTGTTCA
- Phenotype
-
MIM: 611132
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RBKS
PubMed Links
Gene Details
- Gene
- RBKS
- Gene Name
- ribokinase
View Full Product Details