Product Details
- SNP ID
-
rs12472599
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:215940850 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTAAGTGTATGTGTGTTGTGTGTG[C/T]GCGCGCGTGTGTGTGTGTCCATCAT
- Phenotype
-
MIM: 609207
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MREG
PubMed Links
Gene Details
- Gene
- MREG
- Gene Name
- melanoregulin
View Full Product Details