Product Details

SNP ID
rs10851724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:63124503 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTTTTAACCAGGTAGAGGTGTCA[A/T]CTATTTTTTTCAAACAGAAGGCAAA
Phenotype
MIM: 608440
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LACTB PubMed Links

Gene Details

Gene
LACTB
Gene Name
lactamase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288585.1 Intron NP_001275514.1
NM_032857.4 Intron NP_116246.2
NM_171846.3 Intron NP_741982.1
XM_011521180.1 Intron XP_011519482.1

View Full Product Details