Product Details

SNP ID
rs11053063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:34028076 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGTTCCAAGTAAGTTTGTATTCA[C/T]ATGATCTGAGTAGACATTGTCTCTT
Phenotype
MIM: 603313
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALG10 PubMed Links

Gene Details

Gene
ALG10
Gene Name
ALG10, alpha-1,2-glucosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032834.3 Intron NP_116223.3
Gene
LOC105369727
Gene Name
uncharacterized LOC105369727
There are no transcripts associated with this gene.

View Full Product Details