Product Details

SNP ID
rs4281295
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:226950712 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCACAAAATGAAAACATCTGTG[A/G]AACTGGTTTCATTCATGGGGGAGAA
Phenotype
MIM: 606980
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COQ8A PubMed Links

Gene Details

Gene
COQ8A
Gene Name
coenzyme Q8A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020247.4 Intron NP_064632.2
XM_005273201.1 Intron XP_005273258.1
XM_011544238.1 Intron XP_011542540.1
XM_011544239.2 Intron XP_011542541.1
XM_011544240.2 Intron XP_011542542.1
XM_011544241.2 Intron XP_011542543.1
XM_017001852.1 Intron XP_016857341.1

View Full Product Details