Product Details

SNP ID
rs1879963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:46841635 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAGCCCACCATTTAGCCCTGAAGT[A/G]GCTGTTCTGCCTGCTTCATAATCAC
Phenotype
MIM: 609543
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00293 PubMed Links

Gene Details

Gene
LINC00293
Gene Name
long intergenic non-protein coding RNA 293
There are no transcripts associated with this gene.

View Full Product Details