Product Details

SNP ID
rs11014867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:25937101 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTAAAATAATTGATTTACTTACC[A/G]GTATTTTCAAATTAGTAAATTTTGT
Phenotype
MIM: 606808
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101929073 PubMed Links

Gene Details

Gene
LOC101929073
Gene Name
uncharacterized LOC101929073
There are no transcripts associated with this gene.

Gene
MYO3A
Gene Name
myosin IIIA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017433.4 Intron NP_059129.3
XM_011519498.2 Intron XP_011517800.1
XM_011519499.1 Intron XP_011517801.1
XM_011519500.2 Intron XP_011517802.1
XM_011519502.1 Intron XP_011517804.1
XM_011519503.1 Intron XP_011517805.1
XM_011519504.1 Intron XP_011517806.1
XM_011519505.1 Intron XP_011517807.1
XM_011519506.2 Intron XP_011517808.1
XM_011519508.2 Intron XP_011517810.1
XM_011519510.1 Intron XP_011517812.1
XM_011519511.2 Intron XP_011517813.1
XM_011519512.1 Intron XP_011517814.1
XM_011519513.2 Intron XP_011517815.1

View Full Product Details