Product Details

SNP ID
hCV26232499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15841734 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGTCAAAGATGGCACTGTCTTA[A/G]GAAATCTACTCTTTTCCACACAGAA
Phenotype
MIM: 300629
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP1S2 PubMed Links

Gene Details

Gene
AP1S2
Gene Name
adaptor related protein complex 1 sigma 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272071.1 Intron NP_001259000.1
NM_003916.4 Intron NP_003907.3

View Full Product Details