Product Details

SNP ID
rs60230785
Assay Type
Functionally Tested
NCBI dbSNP Submissions
9
Location
Chr.4:48021308 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAAACTTTAGAATCATTGAGTCA[A/G]AGTCCCTCATTTTACAGATGCGGAA
Phenotype
MIM: 123825
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CNGA1 PubMed Links

Gene Details

Gene
CNGA1
Gene Name
cyclic nucleotide gated channel alpha 1
There are no transcripts associated with this gene.

Gene
NIPAL1
Gene Name
NIPA like domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207330.2 Intron NP_997213.1
XM_017007784.1 Intron XP_016863273.1

View Full Product Details