Product Details

SNP ID
rs36060625
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:23807344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCAACCAGCAAACCGAGAAAGA[A/C]ACAAACACCCCCAAGAAGAAGGAGA
Phenotype
MIM: 602916
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UBE2E1 PubMed Links

Gene Details

Gene
UBE2E1
Gene Name
ubiquitin conjugating enzyme E2 E1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202476.1 304 Intron NP_001189405.1
NM_003341.4 304 Missense Mutation GAA,GAC E25D NP_003332.1
NM_182666.2 304 Missense Mutation GAA,GAC E25D NP_872607.1
XM_005265431.4 304 Intron XP_005265488.1
Gene
UBE2E1-AS1
Gene Name
UBE2E1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details