Product Details

SNP ID
hCV26374430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:9782104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATACCAGCAGCTGGCGCAGGGGAA[C/T]GCCGTGGGGGGCTCGGCGGGGGCAC
Phenotype
MIM: 126453 MIM: 606142
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DRD5 PubMed Links

Gene Details

Gene
DRD5
Gene Name
dopamine receptor D5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000798.4 471 Silent Mutation AAC,AAT N25N NP_000789.1
Gene
SLC2A9
Gene Name
solute carrier family 2 member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001290.1 471 Intron NP_001001290.1
NM_020041.2 471 Intron NP_064425.2

View Full Product Details