Product Details

SNP ID
rs1342086
Assay Type
Functionally tested
NCBI dbSNP Submissions
40
Location
Chr.1:28579054 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGCCCTCTGGTAATGAATAGCTAA[C/G]GGCACACGAGGGAGGCCAAATGTAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SNHG12 PubMed Links

Gene Details

Gene
SNHG12
Gene Name
small nucleolar RNA host gene 12
There are no transcripts associated with this gene.

Gene
SNORA16A
Gene Name
small nucleolar RNA, H/ACA box 16A
There are no transcripts associated with this gene.

Gene
SNORA44
Gene Name
small nucleolar RNA, H/ACA box 44
There are no transcripts associated with this gene.

Gene
SNORA61
Gene Name
small nucleolar RNA, H/ACA box 61
There are no transcripts associated with this gene.

Gene
SNORD99
Gene Name
small nucleolar RNA, C/D box 99
There are no transcripts associated with this gene.

Gene
TRNAU1AP
Gene Name
tRNA selenocysteine 1 associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017846.4 Intron NP_060316.1
XM_017001542.1 Intron XP_016857031.1

View Full Product Details