Product Details

SNP ID
rs1049385
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:106238063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCATTAAAGAACAATACAGCTCA[A/G]AGGGAAGGGAGATACAGCAAACAAT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CKAP4 PubMed Links

Gene Details

Gene
CKAP4
Gene Name
cytoskeleton associated protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006825.3 2854 UTR 3 NP_006816.2

View Full Product Details