Product Details
- SNP ID
-
rs1803791
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:100337144 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGACACGTCCACATCACAGTTGCCC[A/C]CAAACTGCCTGTGCTCCTCGATGGT
- Phenotype
-
MIM: 609911
MIM: 191050
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SLC25A47
PubMed Links
Gene Details
- Gene
- SLC25A47
- Gene Name
- solute carrier family 25 member 47
There are no transcripts associated with this gene.
- Gene
- WARS
- Gene Name
- tryptophanyl-tRNA synthetase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004184.3 |
1527 |
Missense Mutation |
GGG,GTG |
G391V |
NP_004175.2 |
NM_173701.1 |
1527 |
Missense Mutation |
GGG,GTG |
G391V |
NP_776049.1 |
NM_213645.1 |
1527 |
Missense Mutation |
GGG,GTG |
G350V |
NP_998810.1 |
NM_213646.1 |
1527 |
Missense Mutation |
GGG,GTG |
G350V |
NP_998811.1 |
XM_005268044.3 |
1527 |
Missense Mutation |
GGG,GTG |
G391V |
XP_005268101.1 |
XM_006720249.3 |
1527 |
Missense Mutation |
GGG,GTG |
G391V |
XP_006720312.1 |
XM_011537133.2 |
1527 |
Missense Mutation |
GGG,GTG |
G391V |
XP_011535435.1 |
XM_011537135.2 |
1527 |
Missense Mutation |
GGG,GTG |
G391V |
XP_011535437.1 |
XM_011537136.2 |
1527 |
Missense Mutation |
GGG,GTG |
G350V |
XP_011535438.1 |
XM_017021627.1 |
1527 |
Missense Mutation |
GGG,GTG |
G391V |
XP_016877116.1 |
XM_017021628.1 |
1527 |
Missense Mutation |
GGG,GTG |
G391V |
XP_016877117.1 |
XM_017021629.1 |
1527 |
Missense Mutation |
GGG,GTG |
G391V |
XP_016877118.1 |
View Full Product Details