Product Details

SNP ID
rs2658556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18481187 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATTAAGCTTGAGACAACTTTCAT[A/T]CAAGAGGGCTTGAAAGGGCAGCCTG
Phenotype
MIM: 601387
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LDHAL6A PubMed Links

Gene Details

Gene
LDHAL6A
Gene Name
lactate dehydrogenase A like 6A
There are no transcripts associated with this gene.

Gene
TSG101
Gene Name
tumor susceptibility 101
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006292.3 Intron NP_006283.1
XM_005253108.3 Intron XP_005253165.1

View Full Product Details