Product Details

SNP ID
rs1126437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:18027651 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCTCTCCACGCACACTCTCATC[A/G]ATTTCCACGCAGGTCTCCTGGTTGC
Phenotype
MIM: 611566
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRPS1L1 PubMed Links

Gene Details

Gene
PRPS1L1
Gene Name
phosphoribosyl pyrophosphate synthetase 1-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175886.2 213 Silent Mutation ATC,ATT I44I NP_787082.1

View Full Product Details