Product Details

SNP ID
rs4329708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:59642976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGCTGTCGCAAAAGGCTGGTGAT[A/C]CTCACTGATGGAAGATGATAGAGAA
Phenotype
MIM: 614660
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PATL1 PubMed Links

Gene Details

Gene
PATL1
Gene Name
PAT1 homolog 1, processing body mRNA decay factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152716.2 3218 Missense Mutation AGG,AGT R651S NP_689929.2
XM_005273833.4 3218 Missense Mutation AGG,AGT R651S XP_005273890.1
XM_005273834.1 3218 Missense Mutation AGG,AGT R621S XP_005273891.1
XM_011544811.2 3218 Missense Mutation AGG,AGT R432S XP_011543113.1
XM_017017339.1 3218 Missense Mutation AGG,AGT R432S XP_016872828.1

View Full Product Details