Product Details

SNP ID
rs5030609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30328904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTCTCAGATTCCTTTCTTGTCT[A/G]TTAGAAACGTATGTCAAACGAGGAT
Phenotype
MIM: 605700
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HCG17 PubMed Links

Gene Details

Gene
HCG17
Gene Name
HLA complex group 17 (non-protein coding)
There are no transcripts associated with this gene.

Gene
HCG18
Gene Name
HLA complex group 18 (non-protein coding)
There are no transcripts associated with this gene.

Gene
TRIM39
Gene Name
tripartite motif containing 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021253.3 482 UTR 5 NP_067076.2
NM_172016.2 482 UTR 5 NP_742013.1
Gene
TRIM39-RPP21
Gene Name
TRIM39-RPP21 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199119.1 482 Intron NP_001186048.1

View Full Product Details