Product Details

SNP ID
rs4645422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:53795759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAGCCGGGTGATTCGGGCTGTT[C/T]CCATCTGGAAGCTACAGAGCACTGC
Phenotype
MIM: 608195
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC1 PubMed Links

Gene Details

Gene
LRRC1
Gene Name
leucine rich repeat containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018214.4 Intron NP_060684.4
XM_011514726.2 Intron XP_011513028.1
XM_011514727.2 Intron XP_011513029.1
XM_017010997.1 Intron XP_016866486.1

View Full Product Details