Product Details

SNP ID
rs394150
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.16:75529734 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
GGTCGCGCACCAGGTGCACGATGCG[C/T]AGGTTGAGCGCGGGGTCGCTGAGCA
Phenotype
MIM: 604817 MIM: 614949
Polymorphism
C/T, Transition substitution
Allele Nomenclature
CHST5 L217L
Literature Links
CHST5 PubMed Links

Gene Details

Gene
CHST5
Gene Name
carbohydrate sulfotransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024533.4 2046 Silent Mutation CTA,CTG L217L NP_078809.2
Gene
TMEM231
Gene Name
transmembrane protein 231
There are no transcripts associated with this gene.

View Full Product Details