Product Details
- SNP ID
-
rs4421128
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:39106426 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATGGTTCACATTTTGTTTTTGGA[C/T]TGCTTTGGGAATGAGTATGTACTGT
- Phenotype
-
MIM: 602731
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FYB
PubMed Links
Gene Details
- Gene
- FYB
- Gene Name
- FYN binding protein
View Full Product Details