Product Details

SNP ID
rs1803707
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:156742172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTACGGTTCTCAGAGCTAAACTT[C/G]CAAAGCTACAGTCAGCAATTTTTCA
Phenotype
MIM: 600339 MIM: 611836
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HDGF PubMed Links

Gene Details

Gene
HDGF
Gene Name
hepatoma-derived growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126050.1 2108 UTR 3 NP_001119522.1
NM_001126051.1 2108 UTR 3 NP_001119523.1
NM_001319186.1 2108 UTR 3 NP_001306115.1
NM_001319187.1 2108 UTR 3 NP_001306116.1
NM_001319188.1 2108 UTR 3 NP_001306117.1
NM_004494.2 2108 UTR 3 NP_004485.1
XM_011509453.2 2108 UTR 3 XP_011507755.2
XM_011509454.2 2108 UTR 3 XP_011507756.1
XM_011509455.2 2108 UTR 3 XP_011507757.1
Gene
MRPL24
Gene Name
mitochondrial ribosomal protein L24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024540.3 2108 Intron NP_078816.2
NM_145729.2 2108 Intron NP_663781.1
XM_011509981.2 2108 Intron XP_011508283.1
XM_011509982.2 2108 Intron XP_011508284.1
Gene
RRNAD1
Gene Name
ribosomal RNA adenine dimethylase domain containing 1
There are no transcripts associated with this gene.

View Full Product Details