Product Details
- SNP ID
-
rs2777491
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:41443512 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGAAAGCATTTGATCAGAGAAGCA[G/T]CATCAAAGGTTCAAACAAGAGTGAT
- Phenotype
-
MIM: 611633
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RTF1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2730055] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RTF1
- Gene Name
- RTF1 homolog, Paf1/RNA polymerase II complex component
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015138.4 |
|
Intron |
|
|
NP_055953.3 |
View Full Product Details