Product Details
- SNP ID
-
hCV26652786
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:149479377 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTACCATCAGGTTGCTTATTTTTG[G/T]TTTATGTTTTTTATTTGTATGCATG
- Phenotype
-
MIM: 300823
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
IDS
PubMed Links
Gene Details
- Gene
- IDS
- Gene Name
- iduronate 2-sulfatase
There are no transcripts associated with this gene.
View Full Product Details