Product Details

SNP ID
rs1053701
Assay Type
Functionally tested
NCBI dbSNP Submissions
33
Location
Chr.1:150622225 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTCCCCACGCTCACGGCTCCTTT[C/T]TCCCACACTCACTGCCCTTCTTCCC
Phenotype
MIM: 603061
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENSA PubMed Links

Gene Details

Gene
ENSA
Gene Name
endosulfine alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004436.2 1001 UTR 3 NP_004427.1
NM_207042.1 1001 UTR 3 NP_996925.1
NM_207043.1 1001 Intron NP_996926.1
NM_207044.1 1001 Intron NP_996927.1
NM_207045.1 1001 UTR 3 NP_996928.1
NM_207046.1 1001 UTR 3 NP_996929.1
NM_207047.1 1001 Intron NP_996930.1
NM_207168.1 1001 Intron NP_997051.1

View Full Product Details