Product Details

SNP ID
rs7413162
Assay Type
Functionally tested
NCBI dbSNP Submissions
61
Location
Chr.1:115701256 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAGTTGGGCTATTCATCATCATC[A/G]TCATCACTGTCATCATTATCCTCTT
Phenotype
MIM: 114251 MIM: 610132
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASQ2 PubMed Links

Gene Details

Gene
CASQ2
Gene Name
calsequestrin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001232.3 1449 Silent Mutation GAC,GAT D395D NP_001223.2
Gene
VANGL1
Gene Name
VANGL planar cell polarity protein 1
There are no transcripts associated with this gene.

View Full Product Details