Product Details
- SNP ID
-
rs4439559
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:48568643 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGACAGCTGACAAGTATTACCCA[C/G]AAAGGCACTGAGTTCCCCTAACCCC
- Phenotype
-
MIM: 149750
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LALBA
PubMed Links
Gene Details
- Gene
- LALBA
- Gene Name
- lactalbumin alpha
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002289.2 |
|
Intron |
|
|
NP_002280.1 |
View Full Product Details