Product Details

SNP ID
rs3913291
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:8813168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTCAGAAAGCCAGATTTTTTTAT[T/C]TAGGTTATCATTAGGAATTCTTTCC
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
TTTY11 PubMed Links

Gene Details

Gene
TTTY11
Gene Name
testis-specific transcript, Y-linked 11 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details