Product Details

SNP ID
rs1857786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:48945872 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCTCTATATGCAAGAAGACAACAC[A/G]GCAGAGAAACCTCAAAACTGACATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM51CP PubMed Links

Gene Details

Gene
TRIM51CP
Gene Name
tripartite motif-containing 51C, pseudogene
There are no transcripts associated with this gene.

View Full Product Details