Product Details

SNP ID
rs3007419
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:6468242 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGTGGGGCGGGCAGCTCAACAGC[C/T]GGACAGGGGTGCGGCGGCGGAGACG
Phenotype
MIM: 606351 MIM: 611101 MIM: 603366
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESPN PubMed Links

Gene Details

Gene
ESPN
Gene Name
espin
There are no transcripts associated with this gene.

Gene
PLEKHG5
Gene Name
pleckstrin homology and RhoGEF domain containing G5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042663.1 2746 Missense Mutation CAG,CGG Q921R NP_001036128.1
NM_001042664.1 2746 Missense Mutation CAG,CGG Q865R NP_001036129.1
NM_001042665.1 2746 Missense Mutation CAG,CGG Q865R NP_001036130.1
NM_001265592.1 2746 Missense Mutation CAG,CGG Q944R NP_001252521.1
NM_001265593.1 2746 Missense Mutation CAG,CGG Q934R NP_001252522.1
NM_001265594.1 2746 Missense Mutation CAG,CGG Q865R NP_001252523.1
NM_020631.4 2746 Missense Mutation CAG,CGG Q865R NP_065682.2
NM_198681.3 2746 Missense Mutation CAG,CGG Q942R NP_941374.2
Gene
TNFRSF25
Gene Name
TNF receptor superfamily member 25
There are no transcripts associated with this gene.

View Full Product Details