Product Details

SNP ID
rs3018328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61625069 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACCTGGAAGGAAGGTTCTGCAGC[A/G]AGTTCATGAGAGACGATATGTGGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPLP0P2 PubMed Links

Gene Details

Gene
RPLP0P2
Gene Name
ribosomal protein lateral stalk subunit P0 pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details