Product Details

SNP ID
rs11864064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:61644505 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCACCTTTGTATGTTCCATCTCTC[G/T]CCATAATGACTGATGGAATTCTTAA
Phenotype
MIM: 603008
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CDH8 PubMed Links

Gene Details

Gene
CDH8
Gene Name
cadherin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001796.4 6049 Intron NP_001787.2
XM_005255760.3 6049 UTR 3 XP_005255817.1
XM_017022829.1 6049 Intron XP_016878318.1

View Full Product Details