Product Details

SNP ID
rs4504399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176317364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGACTAGTTTCTACAAGACTTCA[G/A]AAATATACTTGTCTCCAGGTTCCCT
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
SIMC1 PubMed Links

Gene Details

Gene
SIMC1
Gene Name
SUMO interacting motifs containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308195.1 Intron NP_001295124.1
NM_001308196.1 Intron NP_001295125.1
NM_001308200.1 Intron NP_001295129.1
NM_198567.5 Intron NP_940969.3

View Full Product Details