Product Details

SNP ID
rs2150027
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:170535285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTACTACTGTCATATTTTATTT[C/T]TTAAACTTTCTTTTTAATCATATAG
Phenotype
MIM: 607983
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GORAB PubMed Links

Gene Details

Gene
GORAB
Gene Name
golgin, RAB6 interacting
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146039.1 Intron NP_001139511.1
NM_001320252.1 Intron NP_001307181.1
NM_152281.2 Intron NP_689494.2
XM_006711628.3 Intron XP_006711691.1
XM_011510149.2 Intron XP_011508451.1
XM_011510150.2 Intron XP_011508452.1
XM_011510151.1 Intron XP_011508453.1
XM_017002807.1 Intron XP_016858296.1
Gene
LOC101928650
Gene Name
uncharacterized LOC101928650
There are no transcripts associated with this gene.

View Full Product Details