Product Details
- SNP ID
-
rs1132478
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:204190432 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CGTCCCCGCCCCCCGCCCCCGCCCC[A/G]CATGCTCTGACTCCTTTGGGGTCTG
- Phenotype
-
MIM: 603286
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GOLT1A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs200451423] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GOLT1A
- Gene Name
- golgi transport 1A
There are no transcripts associated with this gene.
- Gene
- KISS1
- Gene Name
- KiSS-1 metastasis-suppressor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002256.3 |
623 |
UTR 3 |
|
|
NP_002247.3 |
View Full Product Details