Product Details
- SNP ID
-
rs4481859
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
57
- Location
-
Chr.1:222559314 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTAGAAACGGTCAAACATTATCTAT[G/T]CATTCTAGCCATGTAGAGAGCAGCA
- Phenotype
-
MIM: 604903
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TAF1A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80291548] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TAF1A
- Gene Name
- TATA-box binding protein associated factor, RNA polymerase I subunit A
View Full Product Details