Product Details

SNP ID
rs4848209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:111869061 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAGTAAACATCTGCCATTTATTGC[A/G]TACTTAATACCTACTAGGCCCTGTG
Phenotype
MIM: 608473
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANAPC1 PubMed Links

Gene Details

Gene
ANAPC1
Gene Name
anaphase promoting complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022662.3 Intron NP_073153.1
XM_011511633.2 Intron XP_011509935.1
XM_011511634.2 Intron XP_011509936.1
XM_011511636.2 Intron XP_011509938.1
XM_011511637.2 Intron XP_011509939.1
XM_017004710.1 Intron XP_016860199.1
XM_017004711.1 Intron XP_016860200.1
XM_017004712.1 Intron XP_016860201.1
XM_017004713.1 Intron XP_016860202.1
XM_017004714.1 Intron XP_016860203.1
XM_017004715.1 Intron XP_016860204.1

View Full Product Details